missing translation for 'onlineSavingsMsg'
Learn More

Myozenin 2 Antibody (1D4), Novus Biologicals™

Product Code. 18373339 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373339 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373339 Supplier Novus Biologicals Supplier No. H00051778M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myozenin 2 Monoclonal antibody specifically detects Myozenin 2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myozenin 2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1D4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05195
Gene Alias C4orf5, calcineurin-binding protein calsarcin-1, Calsarcin-1, chromosome 4 open reading frame 5, CS-1, FATZ related protein 2, FATZ-related protein 2, muscle-specific protein, myozenin 2, myozenin-2
Host Species Mouse
Immunogen MYOZ2 (AAH05195, 1 a.a. ~ 264 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLSHNTMMKQRKQQATAIMKEVHGNDVDGMDLGKRVSIPRDIMLEELSHLSNRGARLFKMRQRRSDKYTFENFQYQSRAQINHSIAMQNGKVDGSNLEGGSQQAPLTPPNTPDPRSPPNPDNIAPGYSGPLKEIPPEKFNTTAVPKYYQSPWEQAISNDPELLEALYPKLFKPEGKAELPDYRSFNRVATPFGGFEKASRMVKFKVPDFELLLLTDPRFMSFVNPLSGRRSFNRTPKGWISENIPIVITTEPTDDTTVPESEDL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51778
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.