missing translation for 'onlineSavingsMsg'
Learn More

Myozenin 1 Antibody (OTI4C2), DyLight 594, Novus Biologicals™

Product Code. 18427854 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427854 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427854 Supplier Novus Biologicals Supplier No. NBP272865DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myozenin 1 Monoclonal antibody specifically detects Myozenin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myozenin 1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4C2
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias calsarcin-2, CS-2, FATZ, Filamin-, actinin- and telethonin-binding protein, MYOZ, myozenin, myozenin 1, myozenin-1, Protein FATZ
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 106-299 of human Myozenin 1 (NP_067068) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 58529
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.