missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOZAP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76547-0.025mg
This item is not returnable.
View return policy
Description
MYOZAP Polyclonal specifically detects MYOZAP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MYOZAP | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| GCOM1, Gup, Gup1, Gup2, myocardial zonula adherens protein, MYOZAP, MYZAP | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 100820829 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| NP_001018110 | |
| MYZAP | |
| Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human MYOZAP. The immunogen is located within amino acids 400 - 450 of MYOZAP. Amino Acid Squence: RLDYLTETQAKTEVETRE | |
| 0.025 mg | |
| Primary | |
| Multiple isoforms of MYOZAP are known to exist. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction