missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYOZ2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100630
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Myozenin2.
The calcineurin-binding protein Myozenin 2, also designated calsarcin-1, is a member of the calsarcin protein family. Calcineurin is a calcium- and calmodulin-dependent protein phosphatase that is involved in controlling the slow fiber gene expression in skeletal muscle and hypertrophy of cardiac muscle. The calsarcins are sarcomeric proteins that couple calcineurin and muscle activity. In cardiac and skeletal muscle cells, Myozenin 2 binds calcineurin to α-actinin at the Z-line of the sarcomere. During embryogenesis, Myozenin 1 and 2 are expressed in developing muscle. The Myozenin 2 gene maps to chromosome 4q and is expressed specifically in adult cardiac and slow-twitch skeletal muscle, while Myozenin 1 is only detected in fast skeletal muscle.
Specifications
| MYOZ2 | |
| Polyclonal | |
| Unconjugated | |
| MYOZ2 | |
| 1110012I24Rik; C4orf5; calcineurin-binding protein calsarcin-1; calsarcin-1; CMH16; CS-1; Fatz-2; FATZ-related protein 2; muscle-specific protein; myoz2; myozenin 2; myozenin-2; myozenin-like 2; Myozl2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 295426, 51778, 59006 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9JJW5, Q9NPC6 | |
| MYOZ2 | |
| A synthesized peptide derived from human MYOZ2(Accession Q9NPC6), corresponding to amino acid residues E71-D121. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction