missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYOZ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100629
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MYOZ1.
Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G.
Specifications
| MYOZ1 | |
| Polyclonal | |
| Unconjugated | |
| MYOZ1 | |
| 2310001N11Rik; alpha-actinin binding protein; AV090278; calcineurin-2; calsarcin 2; calsarcin-2; CS-2; FATZ; filamin-, actinin- and telethonin-binding protein; MYOZ; myoz1; myozenin 1; myozenin precursor; myozenin-1; Protein FATZ; RGD1561064; skeletal muscle-specific protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 498440, 58529, 59011 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9JK37, Q9NP98 | |
| MYOZ1 | |
| A synthesized peptide derived from human MYOZ1(Accession Q9NP98), corresponding to amino acid residues T239-G289. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction