missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myotubularin Related Protein 10 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83246-0.1ML
This item is not returnable.
View return policy
Description
Myotubularin Related Protein 10 Polyclonal specifically detects Myotubularin Related Protein 10 in Human samples. It is validated for Western Blot.
Specifications
| Myotubularin Related Protein 10 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 54893 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| MTMR10, Myotubularin-Related Protein 10 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human Myotubularin Related Protein 10 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction