missing translation for 'onlineSavingsMsg'
Learn More

Myotilin Antibody (OTI8A7) [Alexa Fluor™ 532], Novus Biologicals™

Product Code. 30689042 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689042 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689042 Supplier Novus Biologicals Supplier No. NBP344972AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myotilin Monoclonal antibody specifically detects Myotilin in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myotilin
Applications Western Blot
Classification Monoclonal
Clone OTI8A7
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias 57 kDa cytoskeletal protein, LGMD1A, limb-girdle muscular dystrophy 1A (autosomal dominant), Myofibrillar titin-like Ig domains protein, myotilin, Titin immunoglobulin domain protein, titin immunoglobulin domain protein (myotilin), TTIDLGMD1
Host Species Mouse
Immunogen Full length human recombinant protein of human MYOT (NP_006781) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9499
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.