missing translation for 'onlineSavingsMsg'
Learn More
Learn More
myosin X Antibody (CL9025), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88927
This item is not returnable.
View return policy
Description
myosin X Monoclonal specifically detects myosin X in Human samples. It is validated for Western Blot.
Specifications
| myosin X | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 4651 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| CL9025 | |
| Western Blot 1 ug/ml | |
| FLJ10639, FLJ21066, FLJ22268, KIAA0799FLJ43256, MGC131988, myosin X, myosin-X, Unconventional myosin-10 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: DDAFKDSPNPSEHGHSDQR | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction