Learn More
myosin X Antibody (CL8994) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88926AF532
Product Code. 30308397
  • £558.00 / 0.10mL
Description
myosin X Monoclonal antibody specifically detects myosin X in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
myosin X
Monoclonal
Alexa Fluor 532
FLJ10639, FLJ21066, FLJ22268, KIAA0799FLJ43256, MGC131988, myosin X, myosin-X, Unconventional myosin-10
This antibody was developed using a synthetic peptide derived from Q9HD67, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cytoskeleton Markers, Signal Transduction
4651
Store at 4⪚C in the dark.
IgG1
Immunocytochemistry/Immunofluorescence
CL8994
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified