missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Myosin VIIb Polyclonal antibody specifically detects Myosin VIIb in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry
Specifications
Specifications
| Antigen | Myosin VIIb |
| Applications | ELISA, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp686A08248, myosin VIIB |
| Host Species | Rabbit |
| Immunogen | myosin VIIB. (Uniprot# Q6PIF6) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?