missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin VIIb Antibody [Alexa Fluor« 350], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-27882AF350
This item is not returnable.
View return policy
Description
Myosin VIIb Polyclonal antibody specifically detects Myosin VIIb in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Myosin VIIb | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 4648 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 350 | |
| DKFZp686A08248, myosin VIIB | |
| Myosin VIIb antibody was raised against a peptide corresponding to 15 amino acids near the center of human Myosin VIIb. The immunogen is located within amino acids 1100-1150 of Myosin VIIb. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction