missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin VIIa Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £364.00
Specifications
| Antigen | Myosin VIIa |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Myosin VIIa Polyclonal antibody specifically detects Myosin VIIa in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Myosin VIIa | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 4647 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| deafness, autosomal dominant 11, deafness, autosomal recessive 2, DFNA11, DFNB2, myosin VIIA, myosin-VIIa, MYOVIIA, MYU7A, USH1Bsevere)) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 850-1150 of human MYO7A (NP_000251.3). MIARRLHQRLRAEYLWRLEAEKMRLAEEEKLRKEMSAKKAKEEAERKHQERLAQLAREDAERELKEKEAARRKKELLEQMERARHEPVNHSDMVDKMFGFLGTSGGLPGQEGQAPSGFEDLERGRREMVEEDLDAALPLPDEDEEDLSEYKFAKFAATYFQGTTTHSYTRRPLKQPLLYHDDEGDQLAALAVWITILRFMGDLPEPKYHTAMSDGSEKIPVMTKIYETLGKKTYKRELQALQGEGEAQLPEGQKKSSVRHKLVHLTLKKKSKLTEEVTKRLHDGESTVQGNSMLEDRPTSN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title