missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myosin Skeletal/Smooth Muscle Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA128038
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Myosin proteins are motor proteins critical for muscle contraction in both skeletal and smooth muscle. They convert chemical energy from ATP hydrolysis into mechanical energy, generating force and movement along actin filaments. In skeletal muscle, myosin molecules are part of the contractile apparatus, where they form thick filaments that interact with actin thin filaments to produce contraction. Myosin heavy chain (MyHC) isoforms are key determinants of muscle fiber types, contributing to the diversity in muscle function and specialization. Skeletal muscle fibers can express different MyHC isoforms, resulting in fibers with varying speed and endurance characteristics. Smooth muscle myosin, in contrast, is involved in the involuntary contractions of smooth muscle tissues found in the walls of hollow organs, such as the intestines and blood vessels. Smooth muscle myosin's regulatory mechanisms differ significantly from skeletal muscle, involving phosphorylation of the myosin light chain to initiate contraction.
Specifications
| Myosin Skeletal/Smooth Muscle | |
| Polyclonal | |
| Unconjugated | |
| MYH11 | |
| A530084A17Rik; AAT4; AV071570; contractile protein; epididymis luminal protein 71; FAA4; HEL71; IId; IId/x; KIAA0866; MdMs; MHC2X/D; MHC-2X/D; MYH1; MYH11; myh11 {ECO:0000312; MYHa; MYHC; MyHC-2x; MyHC-2X/D; MyHC-IId/x; MYHC-IIX; myHC-IIx/d; MYHSA1; Myhs-f; Myhsf2; Myhs-f2; myosin heavy chain 1; Myosin heavy chain 11; myosin heavy chain 11, smooth muscle; myosin heavy chain 21; myosin heavy chain 2x; Myosin heavy chain IIx/d; myosin heavy chain isoform SMA; myosin heavy chain SMA; myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain, smooth muscle isoform; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 11, smooth muscle; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 11, smooth muscle; myosin-1; Myosin-11; RGD:3136}; SM1; SM2; SMHC; smMHC; smooth muscle myosin heavy chain; type 2X myosin heavy chain | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Bovine | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with 15mM sodium azide | |
| P12882, P35749, Q9BE40 | |
| MYH1, MYH11 | |
| Whole myosin (heavy and light chains) from bovine uterus. | |
| RUO | |
| 281337, 4619, 4629, 530050 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction