missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Myosin Phosphatase Polyclonal antibody specifically detects Myosin Phosphatase in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Myosin Phosphatase |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 50% glycerol, pH7.3 |
| Gene Alias | MBSmyosin phosphatase, target subunit 1, MGC133042, Myosin phosphatase target subunit 1, Myosin phosphatase-targeting subunit 1, MYPT1protein phosphatase 1 regulatory subunit 12A, protein phosphatase 1, regulatory (inhibitor) subunit 12A, Protein phosphatase myosin-binding subunit |
| Host Species | Rabbit |
| Immunogen | A synthetic phosphorylated peptide around T696 of human PPP1R12A/PPP1R12B/PPP1R12C (NP_002471.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?