missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin light chain kinase Rabbit anti-Human, Mouse, Rat, Clone: 1J2U2, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Myosin light chain kinase |
|---|---|
| Clone | 1J2U2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18313535
|
Novus Biologicals
NBP3-16282-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18342365
|
Novus Biologicals
NBP3-16282-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myosin light chain kinase Monoclonal antibody specifically detects Myosin light chain kinase in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Myosin light chain kinase | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4638 | |
| IgG | |
| Affinity purified |
| 1J2U2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp686I10125, EC 2.7.11, EC 2.7.11.18, FLJ12216, Kinase-related protein, KRP, light polypeptide kinase, MLCK1, MLCK108, MLCKMLCK210, MSTP083, MYLK1, myosin light chain kinase, smooth muscle, smooth muscle myosin light chain kinase, telokin | |
| A synthetic peptide corresponding to a sequence within amino acids 1800-1900 of human Myosin light chain kinase (Q15746). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title