missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin light chain kinase Antibody (28C5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26122-100ul
This item is not returnable.
View return policy
Description
Myosin light chain kinase Monoclonal antibody specifically detects Myosin light chain kinase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| Myosin light chain kinase | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 28C5 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
| DKFZp686I10125, EC 2.7.11, EC 2.7.11.18, FLJ12216, Kinase-related protein, KRP, light polypeptide kinase, MLCK1, MLCK108, MLCKMLCK210, MSTP083, MYLK1, myosin light chain kinase, smooth muscle, smooth muscle myosin light chain kinase, telokin | |
| A synthesized peptide derived from Human Myosin light chain kinase [UniProt Q15746] | |
| 100 μL | |
| Protein Kinase | |
| 4638 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction