missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Myosin Light Chain 2 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10906-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.Tekniske data
| Myosin Light Chain 2 | |
| Polyclonal | |
| Unconjugated | |
| MYL2 | |
| CMH10, MLC 2, MLC 2v, MLC2, MLC2V, MYL2, Myosin Light Chain 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17906, 363925, 4633, 677750 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P08733, P10916, P51667 | |
| Myl2, myl2b | |
| Myosin Light Chain 2 Fusion Protein Ag1356 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human, Zebrafish | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur