missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Light Chain 2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5A1F1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60229-1-IG-150UL
This item is not returnable.
View return policy
Description
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.Specifications
| Myosin Light Chain 2 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08733, P10916, P51667 | |
| Myl2 | |
| Myosin Light Chain 2 Fusion Protein Ag1356 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 5A1F1 | |
| Unconjugated | |
| MYL2 | |
| MLC 2, MLC 2v, MLC2, MLC2v, MYL2, Myosin Light Chain 2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 17906, 363925, 396690, 4633 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction