missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Light Chain 2/MLC-2V Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Shop All Proteintech ProductsDescription
The Myosin Light Chain 2/MLC-2V antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human Myosin Light Chain 2/MLC-2V. This antibody recognizes human, mouse, rat, zebrafish antigen. The Myosin Light Chain 2/MLC-2V antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
| Antigen | Myosin Light Chain 2/MLC-2V |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraLite 488 |
| Host Species | Rabbit |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 4633 |
| Target Species | Human, Mouse, Rat, Zebrafish |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?