missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Light Chain 2 Antibody (F36.5B9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-89029
This item is not returnable.
View return policy
Description
Myosin Light Chain 2 Monoclonal specifically detects Myosin Light Chain 2 in Human samples. It is validated for Western Blot, Immunohistochemistry-Frozen.
Specifications
| Myosin Light Chain 2 | |
| Monoclonal | |
| Unconjugated | |
| cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2 | |
| Human ventricular myosin. | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Immunohistochemistry (Frozen) | |
| F36.5B9 | |
| Sterile filtered supernatant | |
| Mouse | |
| 1 mL | |
| Phospho Specific | |
| 4633 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction