missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Light Chain 2 Antibody (3B2), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-30249APCCY7
This item is not returnable.
View return policy
Description
Myosin Light Chain 2 Monoclonal specifically detects Myosin Light Chain 2 in Human, Mouse, Xenopus samples. It is validated for Flow Cytometry.
Specifications
| Myosin Light Chain 2 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| MYL2 | |
| Recombinant human MYL2 (1-166aa) purified from E. coli (NP_000423). | |
| 0.1 mL | |
| Primary | |
| This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human MYL2 protein. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 3B2 | |
| Flow Cytometry | |
| cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2 | |
| Mouse | |
| Protein G purified | |
| Phospho Specific | |
| 4633.0 | |
| Human, Mouse, Xenopus species | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction