missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Light Chain 2 Antibody (1F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43002
This item is not returnable.
View return policy
Description
Myosin Light Chain 2 Monoclonal antibody specifically detects Myosin Light Chain 2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Myosin Light Chain 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 1F7 | |
| Western Blot 1:1000 - 1:5000, ELISA, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:500 | |
| cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2 | |
| myosin, light chain 2, regulatory, cardiac, slow - (Uniprot# P10916) | |
| 100 μg | |
| Phospho Specific | |
| 4633 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction