missing translation for 'onlineSavingsMsg'
Learn More

Myosin IXA Antibody (4C11), Novus Biologicals™

Product Code. 18354158 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354158 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354158 Supplier Novus Biologicals Supplier No. H00004649M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myosin IXA Monoclonal antibody specifically detects Myosin IXA in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin IXA
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4C11
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_008832
Gene Alias FLJ11061, FLJ13244, MGC71859, MYO9A variant protein, myosin IXA, myosin-IXa, MYR7, Unconventional myosin-9a
Host Species Mouse
Immunogen MYO9A (NP_008832, 719 a.a. ~ 828 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GKRNIHRKTGHDDTAPCAILKSMDSFSFLQHPVHQRSLEILQRCKEEKYSITRKNPRTPLSDLQGMNALNEKNQHDTFDIAWNGRTGIRQSRLSSGTSLLDKDGIFANST
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4649
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.