missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Myosin Heavy Chain Recombinant Rabbit Monoclonal Antibody (6V0G1)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA535613

Product Code. 17126273

  • £437.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Myosin Heavy Chain Recombinant Monoclonal Antibody for ELISA, IHC (P), Western Blot

The function of Mhc is muscle contraction.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Myosin Heavy Chain
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P02563, Q02566, Q5SX40
MYH1, Myh6
Recombinant protein (or fragment).
100 μL
Primary
Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot
6V0G1
Unconjugated
Myh6
A530084A17Rik; A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-MHC; ASD3; beta-myosin heavy chain; Bsh; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; CG17927; CG17927-PA; CG17927-PB; CG17927-PC; CG17927-PD; CG17927-PE; CG17927-PF; CG17927-PG; CG17927-PH; CG17927-PI; CG17927-PK; CG17927-PL; CG17927-PM; CG17927-PN; CG17927-PO; CG17927-PP; CG17927-PQ; CG17927-PR; CG17927-PS; CG17927-PT; CG17927-PU; CG17927-PV; chain; chr2L:16765388..16765564; chr2L:16781880..16782040; CMD1EE; CMH14; DFNA4; DFNA4A; Dm II; Dmel\CG17927; Dmel_CG17927; DmMHC; DROMHC; DroMII; epididymis luminal protein 71; FP17425; heavy chain; HEL71; ifm(2)2; ifm(2)RU1; ifm(2)RU2; IId; IId/x; indirect flight muscle (2) RU1; indirect flight muscle (2) RU2; KIAA2034; l(2)36Ae; l(2)k10423; l(2)M66; MdMs; mhc; MHC16; MHC2X/D; MHC-2X/D; Mhc36B; Mhc-PA; Mhc-PB; Mhc-PC; Mhc-PD; Mhc-PE; Mhc-PF; Mhc-PG; Mhc-PH; Mhc-PI; Mhc-PK; Mhc-PL; Mhc-PM; Mhc-PN; Mhc-PO; Mhc-PP; Mhc-PQ; Mhc-PR; Mhc-PS; Mhc-PT; Mhc-PU; Mhc-PV; mMHC; MRP; muscle myosin; muscle myosin heavy chain; muscle myosin II; muscle myosin II heavy chain; muscle myosin-II; muscle specific myosin II; MYH1; MYH14; MYH14 variant protein; MYH17; MYH6; MYHa; MYHC; MyHC-2x; MyHC-2X/D; Myhca; Myhc-a; MyHC-alpha; MyHC-IId/x; MYHC-IIX; myHC-IIx/d; MYHSA1; Myhs-f; Myhsf2; Myhs-f2; Myo; Myo6; myosin; myosin HC; myosin heavy; myosin heavy chain; myosin heavy chain 1; Myosin heavy chain 14; Myosin heavy chain 2x; myosin heavy chain 6; Myosin heavy chain IIx/d; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; Myosin heavy chain, muscle; Myosin heavy chain, non-muscle IIc; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin heavy chain, skeletal muscle, adult 1; myosin heavy Chain-PF; myosin heavy-chain; myosin heavy-chain protein; myosin II heavy chain; myosin rod protein; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 14, non-muscle; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 14; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin-1; Myosin-14; myosin-6; myosin-heavy chain; NMHC II-C; NMHC-II-C; non-muscle myosin heavy chain IIc; nonmuscle myosin heavy chain II-C; Not-upheld; Nup; PNMHH; samba; sarcomeric myosin heavy-chain; shrunken thorax; Shrunken-thorax; Sht; sMHC; SSS3; Stp; stuckup; type 2X myosin heavy chain
Rabbit
Affinity chromatography
RUO
17879, 17888, 287408, 29556
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.