missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 2 Antibody (TH81 (BGN/04/4481)), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65675IR
This item is not returnable.
View return policy
Description
Myosin heavy chain 2 Monoclonal specifically detects Myosin heavy chain 2 in Human, Rat, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Myosin heavy chain 2 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MYH2 | |
| Myosin heavy chain from rabbit muscle | |
| 0.1 mL | |
| Cytoskeleton Markers | |
| 4620 | |
| Human, Rat, Rabbit | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| TH81 (BGN/04/4481) | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| fast 2a myosin heavy chain, MYH2A, MYHas8, MyHC-2A, MyHC-IIa, MYHSA2, myosin heavy chain 2, myosin heavy chain 2a, myosin heavy chain IIa, myosin, heavy chain 2, skeletal muscle, adult, myosin, heavy polypeptide 2, skeletal muscle, adult, myosin-2, skeletal muscle, adult 2, type IIA myosin heavy chain | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| NB100-65675 reacts strongly with human cardiac myosin and also recognizes rabbit myosin heavy and light chains. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction