missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 2 Antibody (4G9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43001
This item is not returnable.
View return policy
Description
Myosin heavy chain 2 Monoclonal antibody specifically detects Myosin heavy chain 2 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| Myosin heavy chain 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 4G9 | |
| Western Blot 1:5000 - 1:10000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| fast 2a myosin heavy chain, MYH2A, MYHas8, MyHC-2A, MyHC-IIa, MYHSA2, myosin heavy chain 2, myosin heavy chain 2a, myosin heavy chain IIa, myosin, heavy chain 2, skeletal muscle, adult, myosin, heavy polypeptide 2, skeletal muscle, adult, myosin-2, skeletal muscle, adult 2, type IIA myosin heavy chain | |
| myosin, heavy chain 2, skeletal muscle, adult - (Uniprot# Q9UKX2) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 4620 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction