missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £484.00
Specifications
| Antigen | Myosin heavy chain 14 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669236
|
Novus Biologicals
NBP2-49533-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669987
|
Novus Biologicals
NBP2-49533 |
0.1 mL |
£484.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myosin heavy chain 14 Polyclonal antibody specifically detects Myosin heavy chain 14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Myosin heavy chain 14 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers, Hypoxia, Stem Cell Markers | |
| PBS (pH 7.2), 40% Glycerol | |
| 79784 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DFNA4, DFNA4A, FP17425, MHC16, MYH14, MYH17, myosin, heavy chain 14, non-muscle, NMHC II-C, NMHC-II-C, PNMHH | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FTTRTVRQVFRLEEGVASDEEAEEAQPGSGPSPEPEGSPP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title