missing translation for 'onlineSavingsMsg'
Learn More

Myosin heavy chain 14 Antibody [HRP], Novus Biologicals™

Product Code. 30761093 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30761093 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30761093 Supplier Novus Biologicals Supplier No. NBP346983H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Myosin heavy chain 14 Polyclonal antibody specifically detects Myosin heavy chain 14 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin heavy chain 14
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias DFNA4, DFNA4A, FP17425, MHC16, MYH14, MYH17, myosin, heavy chain 14, non-muscle, NMHC II-C, NMHC-II-C, PNMHH
Host Species Rabbit
Immunogen myosin, heavy chain 14. (Uniprot# Q7Z406)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Hypoxia, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 79784
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.