missing translation for 'onlineSavingsMsg'
Learn More

Myosin heavy chain 13 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£149.00 - £359.00

Specifications

Antigen Myosin heavy chain 13
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/ Immunofluorescence 1:50 - 1:100
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18650068
View Documents
Novus Biologicals
NBP3-05614-100ul
100 μg
£359.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18615268
View Documents
Novus Biologicals
NBP3-05614-20ul
20 μg
£149.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Myosin heavy chain 13 Polyclonal antibody specifically detects Myosin heavy chain 13 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

Myosin heavy chain 13
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
Mouse, Rat
MYH13, MYH13 myosin, heavy chain 13, skeletal muscle, MyHC-eo
Recombinant fusion protein containing a sequence corresponding to amino acids 1240-1320 of human Myosin heavy chain 13 (NP_003793.2). EALSKSKSNIERTCRTVEDQFSEIKAKDEQQTQLIHDLNMQKARLQTQNGELSHRVEEKESLISQLTKSKQALTQQLEELK
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/ Immunofluorescence 1:50 - 1:100
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
8735
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.