missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 11 Rabbit anti-Human, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Rabbit, Clone: MYH11/2303R, Novus Biologicals™
Rabbit Monoclonal Antibody
£245.00
Specifications
| Antigen | Myosin heavy chain 11 |
|---|---|
| Clone | MYH11/2303R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
Myosin heavy chain 11 Monoclonal specifically detects Myosin heavy chain 11 in Human, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Rabbit samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Myosin heavy chain 11 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Hypoxia, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 4629 | |
| IgG | |
| Protein A or G purified |
| MYH11/2303R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Pig, Bovine, Canine, Chicken, Feline, Guinea Pig, Rabbit | |
| AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain | |
| Recombinant full-length human Myosin heavy chain 11 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title