missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Myosin heavy chain 11 Antibody (CL14617), Novus Biologicals™
Mouse Monoclonal Antibody
£298.00 - £449.00
Specifications
| Antigen | Myosin heavy chain 11 |
|---|---|
| Clone | CL14617 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043299
|
Novus Biologicals™
NBP3-26874-25ul |
25 μL |
£298.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043736
|
Novus Biologicals™
NBP3-26874-100ul |
100 μL |
£449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myosin heavy chain 11 Monoclonal antibody specifically detects Myosin heavy chain 11 in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| Myosin heavy chain 11 | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Hypoxia, Stem Cell Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 4629 | |
| IgG1 | |
| Protein A purified |
| CL14617 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain | |
| This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of P35749. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title