missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Myosin heavy chain 1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5145318

 View more versions of this product

Product Code. 17970273

  • £161.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Myosin is a protein which is known to interact with actin in muscle and non-muscle cells. It contains two identical heavy chains and four light chains. Myosin molecules consist of two major regions: tails (rods) and heads. They aggregate into filaments through the tail region and interact with actin and with ATP through the head region. Multiple forms of myosin heavy chains exist for each muscle type-skeletal, cardiac, smooth and non-muscle isomyosin forms exist in different types of skeletal muscle, depending on the physiological function of the muscle. They are designated at type I (slow twitch) and type II (fast twitch).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Myosin heavy chain 1
Polyclonal
Unconjugated
Myh1
A530084A17Rik; epididymis luminal protein 71; HEL71; IId; IId/x; MdMs; MHC2X/D; MHC-2X/D; MYH1; MYHa; MYHC; myHC-2x; MyHC-2X/D; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; MYHSA1; Myhs-f; Myhsf2; Myhs-f2; myosin heavy chain 1; Myosin heavy chain 2x; myosin heavy chain IIx/d; Myosin heavy chain, skeletal muscle, adult 1; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin-1; type 2X myosin heavy chain
Rabbit
Antigen affinity chromatography, Protein A
RUO
100125538, 17879, 287408, 4619
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
P12882, Q5SX40, Q9TV61
Myh1
Recombinant Human MYH1 protein, Tyr585-Ala794 (Accession #P12882).
20 μL
Primary
Human, Mouse, Pig, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.