missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Heavy Chain 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15029
This item is not returnable.
View return policy
Description
Myosin Heavy Chain 1 Polyclonal specifically detects Myosin Heavy Chain 1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Myosin Heavy Chain 1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7), 20% Glycerol, 1% BSA with 0.025% Proclin 300 | |
| MyHC-2x, MYH1, MYH1 myosin, heavy chain 1, skeletal muscle, adult MYHa, MYHC 1, MYHC1, MYHC-1, MyHC-2X/D, MYHSA1 | |
| Rabbit | |
| 223 kDa | |
| 0.1 mL | |
| Primary | |
| Mouse. Predicted cross-reactivity based sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.92mg/mL | |
| Western Blot 1:500-1:3000 | |
| P12882 | |
| MYH1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human MYH1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 4619 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction