missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Myosin Heavy Chain 1 Polyclonal antibody specifically detects Myosin Heavy Chain 1 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
Specifications
| Antigen | Myosin Heavy Chain 1 |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DFNA4, FLJ13881, FLJ43092, KIAA2034DKFZp667A1311, MHC16, MYH14 variant protein, MYH17, myosin, Myosin heavy chain 14, Myosin heavy chain, non-muscle IIc, myosin, heavy chain 14, myosin, heavy chain 14, non-muscle, myosin, heavy polypeptide 14, myosin-14, NMHC II-C, NMHC-II-C, nonmuscle myosin heavy chain II-C, Non-muscle myosin heavy chain IIc |
| Host Species | Rabbit |
| Immunogen | myosin, heavy chain 1, skeletal muscle, adult. (Uniprot# P12882) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?