Learn More
Myosin 4 Monoclonal Antibody (MF20), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 50-6503-82
324.00 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: This MF20 monoclonal antibody recognizes the heavy chain of myosin II, specificially the light meromyosin portion, in cardiac and skeletal muscle of vertebrates. Myosin II is composed of two heavy chains and four light chains. The 220-kDa myosin heavy chain exists as four different isoforms due to alternative splicing. Myosins interact with actin and hydrolyze ATP to function in muscle contraction, cytokinesis, and phagocytosis. The MF20 has been shown to react to myosin from a variety of mammalian, avian and amphibian species, including rat, mouse, human, chicken, zebrafish, and dog. Applications Reported: This MF20 antibody has been reported for use in immunohistochemical staining, immunocytochemistry, and immunohistochemical staining of frozen tissue sections. Applications Tested: This MF20 antibody has been tested immunocytochemistry on fixed and permeabilized C2C12 cells at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Target protein includes a fast myosin isoform expressed in skeletal muscle and has function involved in Muscle contraction.Specifications
Myosin 4 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q076A5, Q28641, Q29RW1, Q5SX39, Q9Y623 | |
MYH4 | |
Affinity chromatography | |
RUO | |
100009548, 100125991, 100726456, 17884, 360543, 408020, 4622, 468396, 479502 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
MF20 | |
eFluor 660 | |
MYH4 | |
2B myosin heavy chain; AI506973; fast myosin heavy chain 2B; LOW QUALITY PROTEIN: myosin-4; MHC; MHC2B; Minimsc; Minmus; MM; MYH1; MYH2B; MYH-2B; myh4; MyHC IIb isoform; myHC-2b; MyHC-IIb; Myhsf; myosin heavy chain; myosin heavy chain 1; myosin heavy chain 2b; myosin heavy chain 2X; myosin heavy chain 4; Myosin heavy chain IIb; myosin heavy chain IIb isoform; myosin heavy chain isoform 2b; myosin heavy chain type IIb; Myosin heavy chain, skeletal muscle; myosin heavy chain, skeletal muscle, fetal; Myosin heavy chain, skeletal muscle, juvenile; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 4, skeletal muscle; myosin-4; unnamed protein product | |
Mouse | |
100 μg | |
Primary | |
Bovine, Rat, Chicken, Chicken, Feline, Guinea Pig, Guinea Pig, Mouse, Non-human Primate, Non-human Primate, Rabbit | |
Antibody | |
IgG2b κ |
For Research Use Only.