missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Myosin 4 Monoclonal Antibody (MF20), Alexa Fluor™ 488, eBioscience™, Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£180.00 - £445.00

Specifications

Antigen Myosin 4
Clone MF20
Concentration 0.5 mg/mL
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15392050
View Documents
Invitrogen™
53-6503-80
25 μg
£180.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15302060
GREENER_CHOICE
View Documents
Invitrogen™
53-6503-82
100 μg
£445.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: This MF20 monoclonal antibody recognizes the heavy chain of myosin II, specificially the light meromyosin portion, in cardiac and skeletal muscle of vertebrates. Myosin II is composed of two heavy chains and four light chains. The 220-kDa myosin heavy chain exists as four different isoforms due to alternative splicing. Myosins interact with actin and hydrolyze ATP to function in muscle contraction, cytokinesis, and phagocytosis. The MF20 has been shown to react to myosin from a variety of mammalian, avian and amphibian species, including rat, mouse, human, chicken, zebrafish, and dog. Applications Reported: This MF20 antibody has been reported for use in immunohistochemical staining, immunocytochemistry, and immunohistochemical staining of frozen tissue sections. Applications Tested: This MF20 antibody has been tested by immunocytochemistry on fixed and permeabilized C2C12 cells that were differentiated for 2 days in low serum medium prior to staining. Staining can be performed using less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

Target protein includes a fast myosin isoform expressed in skeletal muscle and has function involved in Muscle contraction.
Specifications

Specifications

Myosin 4
0.5 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
2B myosin heavy chain; AI506973; fast myosin heavy chain 2B; LOW QUALITY PROTEIN: myosin-4; MHC; MHC2B; Minimsc; Minmus; MM; MYH1; MYH2B; MYH-2B; myh4; MyHC IIb isoform; myHC-2b; MyHC-IIb; Myhsf; myosin heavy chain; myosin heavy chain 1; myosin heavy chain 2b; myosin heavy chain 2X; myosin heavy chain 4; Myosin heavy chain IIb; myosin heavy chain IIb isoform; myosin heavy chain isoform 2b; myosin heavy chain type IIb; Myosin heavy chain, skeletal muscle; myosin heavy chain, skeletal muscle, fetal; Myosin heavy chain, skeletal muscle, juvenile; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 4, skeletal muscle; myosin-4; unnamed protein product
MYH4
Primary
4°C, store in dark, DO NOT FREEZE!
MYH4
MF20
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Alexa Fluor 488
Mouse
Bovine, Canine, Chicken, Chimpanzee, Feline, Guinea Pig, Human, Mouse, Monkey, Rabbit, Rat
Q076A5, Q28641, Q29RW1, Q5SX39, Q9Y623
100009548, 100125991, 100726456, 17884, 360543, 408020, 4622, 468396, 479502
IgG2b κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.