missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin 1B Antibody (S04-6D4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19708-50ul
This item is not returnable.
View return policy
Description
Myosin 1B Monoclonal antibody specifically detects Myosin 1B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Myosin 1B | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:20-1:100, Immunohistochemistry-Paraffin | |
| MMIa, MMI-alpha, MYH-1c, MYO1B variant protein, Myosin I alpha, myosin IB, myosin-I alpha, myosin-Ib, myr1 | |
| Recombinant protein of human MYO1B (Uniprot # O43795) | |
| 50 μg | |
| Vision | |
| 4430 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S04-6D4 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction