missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin 18B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£222.00
Specifications
| Antigen | Myosin 18B |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Myosin 18B Polyclonal specifically detects Myosin 18B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Myosin 18B | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| BK125H2.1, DKFZp434C2422, DKFZp779C1668, FLJ41942, myosin 18B, myosin XVIIIB, myosin-XVIIIb | |
| MYO18B | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8IUG5 | |
| 84700 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GSDPFSWKLPSLDYERKTKVDFDDFLPAIRKPQTPTSLAGSAKGGQDGSQRSSIHFETEEANRSFLSGIKTILKKSPEPKEDPAHLSDSSSSSGSIVSFKSADSIKSRPGIPRLAGDGGERTSPERREPGTGRKDDDVASIMKKY | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts