missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myoneurin Antibody (OTI3B9), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72864IR
This item is not returnable.
View return policy
Description
Myoneurin Monoclonal specifically detects Myoneurin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Myoneurin | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MYNN | |
| Recombinant protein fragment corresponding to amino acids 10-300 of human Myoneurin (NP_061127) produced in E.coli. | |
| 0.1 mL | |
| 55892.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI3B9 | |
| Western Blot, Flow Cytometry | |
| myoneurin, OSZF, ZBTB31SBBIZ1, Zinc finger and BTB domain-containing protein 31, zinc finger protein with BTB/POZ domain, ZNF902 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction