missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myoneurin Antibody (OTI3B9), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72864APCCY7
This item is not returnable.
View return policy
Description
Myoneurin Monoclonal specifically detects Myoneurin in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Myoneurin | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| MYNN | |
| Recombinant protein fragment corresponding to amino acids 10-300 of human Myoneurin (NP_061127) produced in E.coli. | |
| 0.1 mL | |
| 55892.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI3B9 | |
| Flow Cytometry | |
| myoneurin, OSZF, ZBTB31SBBIZ1, Zinc finger and BTB domain-containing protein 31, zinc finger protein with BTB/POZ domain, ZNF902 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction