missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myomegalin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530324
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, PC-3, SK-N-SH, mouse brain. Predicted reactivity: Human (98%), Mouse (83%), Rat (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene serves to anchor phosphodiesterase 4D to the Golgi/centrosome region of the cell. Defects in this gene may be a cause of myeloproliferative disorder (MBD) associated with eosinophilia. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| Myomegalin | |
| Polyclonal | |
| Unconjugated | |
| PDE4DIP | |
| 4732458A06Rik; 9430063L05Rik; C87016; cardiomyopathy-associated protein 2; CMYA2; D130016K21Rik; D3Bwg1078e; KIAA0454; KIAA0477; mKIAA0454; MMGL; Myomegalin; myomegalin/phosphodiesterase 4D interacting protein variant 8; Pbp46; PDE4DIP; phosphodiesterase 4D interacting protein; phosphodiesterase 4D interacting protein (myomegalin); phosphodiesterase 4D-interacting protein; Phosphodiesterase-binding protein clone 46; upregulated during skeletal muscle growth 4; Usmg4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83679, 9659 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.81 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q5VU43, Q80YT7 | |
| PDE4DIP | |
| Recombinant protein encompassing a sequence within the center region of human Myomegalin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction