missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYOM3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100627
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MYOM3.
MYOM3 (myomesin 3) is a structural component of the M-band in striated muscle and is involved in sarcomere stability and resistance during intense or sustained stretching. MYOM3 can be detected mainly in intermediate speed fibers of skeletal muscle. Recently high level of MYOM3 fragments were detected in sera from patients with muscular dystrophy, including Duchenne muscular dystrophy (DMD). MYOM3 fragments may be used as serum biomarker for DMD and other neuromuscular disorders.
Specifications
| MYOM3 | |
| Polyclonal | |
| Unconjugated | |
| Myom3 | |
| 8430427K15Rik; Gm434; MYOM3; myomesin 3; Myomesin family member 3; myomesin family, member 3; myomesin-3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 127294, 242702 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2ABU4, Q5VTT5 | |
| Myom3 | |
| A synthesized peptide derived from human MYOM3(Accession Q5VTT5), corresponding to amino acid residues N1089-T1139. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction