missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYOM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551017
This item is not returnable.
View return policy
Description
MYOM2 Polyclonal Antibody detects endogenous levels of MYOM2 protein.
The giant protein titin, together with its associated proteins, interconnects the major structure of sarcomeres, the M bands and Z discs. The C-terminal end of the titin string extends into the M line, where it binds tightly to M-band constituents of apparent molecular masses of 190 kD and 165 kD. The predicted MYOM2 protein contains 1,465 amino acids. Like MYOM1, MYOM2 has a unique N-terminal domain followed by 12 repeat domains with strong homology to either fibronectin type III or immunoglobulin C2 domains. Protein sequence comparisons suggested that the MYOM2 protein and bovine M protein are identical. [provided by RefSeq, Jul 2008].
Specifications
| MYOM2 | |
| Polyclonal | |
| Unconjugated | |
| MYOM2 | |
| 165 kDa connectin-associated protein; 165 kDa titin-associated protein; AW146149; M-band protein; M-protein; MYOM2; myomesin (M-protein) 2, 165kDa; myomesin 2; myomesin family member 2; Myomesin-2; skeletal muscle 165kD protein; skeletal muscle 165kDa protein; titin-associated protein, 165 kD; TTNAP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17930, 9172 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| P54296 | |
| MYOM2 | |
| Synthesized peptide derived from the Internal region of human Myomesin-2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction