missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOM2 Polyclonal Antibody

MYOM2 Polyclonal Antibody
Brand: Thermo Scientific PA551632
This item is not returnable.
View return policy
Description
MYOM2 Polyclonal Antibody for ICC/IF, IHC
Specifications
| MYOM2 | |
| Polyclonal | |
| Unconjugated | |
| MYOM2 | |
| 165 kDa connectin-associated protein; 165 kDa titin-associated protein; AW146149; M-band protein; M-protein; MYOM2; myomesin (M-protein) 2, 165kDa; myomesin 2; myomesin family member 2; Myomesin-2; skeletal muscle 165kD protein; skeletal muscle 165kDa protein; titin-associated protein, 165 kD; TTNAP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9172 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P54296 | |
| MYOM2 | |
| Recombinant protein corresponding to Human MYOM2. Recombinant protein control fragment (Product #RP-100909). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction