Learn More
MYOM1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22084-1-AP-150UL
 View more versions of this product
Product Code. 16891625
  • £349.00 / 150µL
Description
The MYOM1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human MYOM1. This antibody recognizes human, mouse, rat antigen. The MYOM1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
MYOM1
Polyclonal
Unconjugated
150 μL
8736
IgG
Western Blot, Immunohistochemistry, ELISA
447 μg/mL
Rabbit
Primary
Human, Rat, Mouse