missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myoglobin Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149503
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS. If 0.5 mL of PBS is used, the antibody concentration will be 0.2 mg/mL.
This gene encodes a member of the globin superfamily and is expressed in skeletal and cardiac muscles. The encoded protein is a hemoprotein contributing to intracellular oxygen storage and transcellular facilitated diffusion of oxygen. At least three alternatively spliced transcript variants encoding the same protein have been reported.
Specifications
| Myoglobin | |
| Polyclonal | |
| Unconjugated | |
| MB | |
| AI325109; Human myoglobin; Mb; MGC13548; myoglobgin; myoglobin; OTTHUMP00000197936; PVALB | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 4151 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and 0.05% sodium azide | |
| P02144 | |
| MB | |
| Human cardiac Myoglobin. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction