missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myoglobin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109945
This item is not returnable.
View return policy
Description
Immunogen sequence: MGLSDGEWQL VLNVWGKVEA DIPGHGQEVL IRLFKGHPET LEKFDKFKHL KSEDEMKASE DLKKHGATVL TALGGILKKK GHHEAEIKPL AQSHATKHKI PVKYLEFISE CIIQVLQSKH PGDFGADAQG AMNKALELFR KDMASNYKEL GFQG.
This gene encodes a member of the globin superfamily and is expressed in skeletal and cardiac muscles. The encoded protein is a hemoprotein contributing to intracellular oxygen storage and transcellular facilitated diffusion of oxygen. At least three alternatively spliced transcript variants encoding the same protein have been reported.
Specifications
| Myoglobin | |
| Polyclonal | |
| Unconjugated | |
| MB | |
| AI325109; Human myoglobin; Mb; MGC13548; myoglobgin; myoglobin; OTTHUMP00000197936; PVALB | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17189, 4151 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.71 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02144, P04247 | |
| MB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-154 of human Myoglobin (NP_0053591). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction