missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myoglobin Antibody (4E2cc), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1291
This item is not returnable.
View return policy
Description
Myoglobin Monoclonal specifically detects Myoglobin in Human samples. It is validated for Immunoassay, Sandwich ELISA.
Specifications
| Myoglobin | |
| Monoclonal | |
| Unconjugated | |
| MGC13548, myoglobin, PVALB | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Myoglobin (4E2) | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| ELISA | |
| 400 | |
| Sandwich ELISA 1:100-1:2000 | |
| MB | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human cardiac myoglobin. | |
| 0.2 mg | |
| Cancer, Cardiovascular Biology | |
| 4151 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction