missing translation for 'onlineSavingsMsg'
Learn More

Myoglobin Antibody (4E2cc), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18433584 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433584 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433584 Supplier Novus Biologicals Supplier No. NB6001291JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myoglobin Monoclonal antibody specifically detects Myoglobin in Human samples. It is validated for Immunoassay, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myoglobin
Applications Immunoassay, ELISA
Classification Monoclonal
Clone 4E2cc
Conjugate Janelia Fluor 549
Dilution Immunoassay, Sandwich ELISA
Formulation 50mM Sodium Borate
Gene Alias MGC13548, myoglobin, PVALB
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human cardiac myoglobin.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 4151
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.