missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myoglobin Antibody (4E2cc) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB600-1291JF525
This item is not returnable.
View return policy
Description
Myoglobin Monoclonal antibody specifically detects Myoglobin in Human samples. It is validated for Immunoassay,Sandwich ELISA
Specifications
| Myoglobin | |
| Monoclonal | |
| Janelia Fluor 525 | |
| MGC13548, myoglobin, PVALB | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human cardiac myoglobin. | |
| 0.2 mL | |
| Cancer, Cardiovascular Biology | |
| 4151 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunoassay, Sandwich ELISA | |
| 4E2cc | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction